N-Methylformamide 99%

Supplier: Apollo Scientific
Danger

Synonyms: Formmethylamide, Formic acid methylamide

OR921948-1KG
APOSOR921948-1KGEA 473 EUR
APOSOR921948-1KG
N-Methylformamide 99%
N-Methylformamide
Formula: C₂H₅NO
MW: 59.07 g/mol
Boiling Pt: 181…185 °C
Melting Pt: –4 °C
Density: 1.018 g/cm³ (25 °C)
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00003280
CAS Number: 123-39-7
EINECS: 204-624-6
Merck Index: 13,00061

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR