5-Cyclopropylisoxazole-4-carboxylic acid 95%

Supplier: Apollo Scientific

OR922620-1G
APOSOR922620-1GEA 156 EUR
APOSOR922620-1G
5-Cyclopropylisoxazole-4-carboxylic acid 95%
5-Cyclopropylisoxazole-4-carboxylic acid
Formula: C₇H₇NO₃
MW: 153.135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798100
CAS Number: 124845-04-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR