2-Chloro-5-cyanobenzene-1-sulphonyl chloride 95%

Supplier: Apollo Scientific
Danger

OR931550-1G OR931550-250MG OR931550-5G
APOSOR931550-1GEA 108 EUR
APOSOR931550-1G APOSOR931550-250MG APOSOR931550-5G
2-Chloro-5-cyanobenzene-1-sulphonyl chloride 95%
2-Chloro-5-cyanobenzene-1-sulphonyl chloride
Formula: C₇H₃Cl₂NO₂S
MW: 236.075 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18392427
CAS Number: 942199-56-6
UN: 1759
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR