1-Bromo-3,3-dimethylbutane 98%

Supplier: Apollo Scientific

OR932225-100G
APOSOR932225-100GEA 120 EUR
APOSOR932225-100G
1-Bromo-3,3-dimethylbutane 98%
1-Bromo-3,3-dimethylbutane
Formula: C₆H₁₃Br
MW: 165.07 g/mol
Boiling Pt: 134…140 °C
Density: 1.1556 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00060844
CAS Number: 1647-23-0
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR