1-(3-Hydroxypyridin-2-yl)ethanone 95%

Supplier: Apollo Scientific

OR932930-1G OR932930-5G
APOSOR932930-1GEA 149 EUR
APOSOR932930-1G APOSOR932930-5G
1-(3-Hydroxypyridin-2-yl)ethanone 95%
1-(3-Hydroxypyridin-2-yl)ethanone
Formula: C₇H₇NO₂
MW: 137.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062669
CAS Number: 13210-29-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR