4-(Dimethylamino)-1,2,2,6,6-pentamethylpiperidine 95%

Supplier: Apollo Scientific

Synonyms: N,N,1,2,2,6,6-Heptamethylpiperidin-4-amine

OR939493-1G
APOSOR939493-1GEA 137 EUR
APOSOR939493-1G
4-(Dimethylamino)-1,2,2,6,6-pentamethylpiperidine 95%
4-(Dimethylamino)-1,2,2,6,6-pentamethylpiperidine
Formula: C₁₂H₂₆N₂
MW: 198.348 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011577
CAS Number: 36177-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR