3-Amino-1-benzofuran-2-carbonitrile 95%

Supplier: Apollo Scientific

OR941653-1G OR941653-250MG OR941653-5G
APOSOR941653-1GEA 121 EUR
APOSOR941653-1G APOSOR941653-250MG APOSOR941653-5G
3-Amino-1-benzofuran-2-carbonitrile 95%
3-Amino-1-benzofuran-2-carbonitrile
Formula: C₉H₆N₂O
MW: 158.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06334923
CAS Number: 62208-67-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR