4-Chloro-2-methyl-6-propylpyrimidine 95%

Supplier: Apollo Scientific

OR944950-1G OR944950-5G
APOSOR944950-1GEA 168 EUR
APOSOR944950-1G APOSOR944950-5G
4-Chloro-2-methyl-6-propylpyrimidine 95%
4-Chloro-2-methyl-6-propylpyrimidine
Formula: C₈H₁₁ClN₂
MW: 170.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07365353
CAS Number: 89967-20-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR