3-Amino-6-cyanopyridazine 97%

Supplier: Apollo Scientific
Warning

OR952513-1G
APOSOR952513-1GEA 453 EUR
APOSOR952513-1G
3-Amino-6-cyanopyridazine 97%
3-Amino-6-cyanopyridazine
Formula: C₅H₄N₄
MW: 120.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743651
CAS Number: 340759-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR