3-(2-Hydroxyphenyl)phenol 96%

Supplier: Apollo Scientific

OR954774-1G
APOSOR954774-1GEA 538 EUR
APOSOR954774-1G
3-(2-Hydroxyphenyl)phenol 96%
3-(2-Hydroxyphenyl)phenol
Formula: C₁₂H₁₀O₂
MW: 186.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00137785
CAS Number: 31835-45-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR