5-Chloro-2-methoxy-3-methylpyridine 95%

Supplier: Apollo Scientific

OR956947-1G
APOSOR956947-1GEA 632 EUR
APOSOR956947-1G
5-Chloro-2-methoxy-3-methylpyridine 95%
5-Chloro-2-methoxy-3-methylpyridine
Formula: C₇H₈ClNO
MW: 157.598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18415742
CAS Number: 1261488-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR