2-(1,1-Dimethylethyl)-4-pyridinamine 95%

Supplier: Apollo Scientific

Synonyms: 2-(tert-Butyl)pyridin-4-amine

OR961242-1G OR961242-250MG
APOSOR961242-1GEA 630 EUR
APOSOR961242-1G APOSOR961242-250MG
2-(1,1-Dimethylethyl)-4-pyridinamine 95%
2-(1,1-Dimethylethyl)-4-pyridinamine
Formula: C₉H₁₄N₂
MW: 150.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01646253
CAS Number: 39919-69-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR