2-Chloro-3,6-difluorobenzyl alcohol 98%

Supplier: Apollo Scientific

PC0003-5G
APOSPC0003-5GEA 79.9 EUR
APOSPC0003-5G
2-Chloro-3,6-difluorobenzyl alcohol 98%
2-Chloro-3,6-difluorobenzyl alcohol
Formula: C₇H₅ClF₂O
MW: 178.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631445
CAS Number: 261762-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR