4-Chloro-2-fluorobenzonitrile 98%

Supplier: Apollo Scientific
Danger

PC0162-1G PC0162-25G PC0162-5G
APOSPC0162-1GEA 14.7 EUR
APOSPC0162-1G APOSPC0162-25G APOSPC0162-5G
4-Chloro-2-fluorobenzonitrile 98%
4-Chloro-2-fluorobenzonitrile
Formula: C₇H₃ClFN
MW: 155.56 g/mol
Boiling Pt: 92 °C/22mmHg
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00143284
CAS Number: 57381-51-8
EINECS: 260-712-4
UN: 3439
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR