4-Bromo-2,6-difluorobenzoic acid

Supplier: Apollo Scientific
Warning

PC0737-100G PC0737-5G PC0737-25G
APOSPC0737-100GEA 210 EUR
APOSPC0737-100G APOSPC0737-5G APOSPC0737-25G
4-Bromo-2,6-difluorobenzoic acid
4-Bromo-2,6-difluorobenzoic acid
Formula: C₇H₃BrF₂O₂
MW: 237 g/mol
Melting Pt: 198…204 °C
Storage Temperature: Ambient
MDL Number: MFCD03094085
CAS Number: 183065-68-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR