Boc-3,4,5-trifluoro-L-phenylalanine

Supplier: Apollo Scientific

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-3-(3,4,5-trifluorophenyl)propanoic acid, 3,4,5-Trifluoro-L-phenylalanine, N-BOC protected

PC1002-1G
APOSPC1002-1GEA 209 EUR
APOSPC1002-1G
Boc-3,4,5-trifluoro-L-phenylalanine
Boc-3,4,5-trifluoro-L-phenylalanine
Formula: C₁₄H₁₆F₃NO₄
MW: 319.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00797558
CAS Number: 205445-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR