N-Allyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine

Supplier: Apollo Scientific

PC10251-1G PC10251-500MG PC10251-5G
APOSPC10251-1GEA 210 EUR
APOSPC10251-1G APOSPC10251-500MG APOSPC10251-5G
N-Allyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine
N-Allyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine
Formula: C₉H₈ClF₃N₂
MW: 236.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00664527
CAS Number: 338761-50-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR