1-(4-Fluoro-3-nitrophenyl)-1H-pyrrole-2,5-dione

Supplier: Apollo Scientific
Warning

Synonyms: N-(4-Fluoro-3-nitrophenyl)maleimide

PC10591-1G
APOSPC10591-1GEA 40.6 EUR
APOSPC10591-1G
1-(4-Fluoro-3-nitrophenyl)-1H-pyrrole-2,5-dione
1-(4-Fluoro-3-nitrophenyl)-1H-pyrrole-2,5-dione
Formula: C₁₀H₅FN₂O₄
MW: 236.16 g/mol
Melting Pt: 127…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00591947
CAS Number: 67154-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR