Potassium (2-methoxyphenyl)trifluoroborate 98%

Supplier: Apollo Scientific
Warning

Synonyms: Potassium trifluoro(2-methoxyphenyl)borate, (2-Methoxyphenyl)trifluoroboric acid potassium salt

PC11222-5G PC11222-25G
APOSPC11222-5GEA 60.9 EUR
APOSPC11222-5G APOSPC11222-25G
Potassium (2-methoxyphenyl)trifluoroborate 98%
Potassium (2-methoxyphenyl)trifluoroborate
Formula: C₇H₇BF₃KO
MW: 214.04 g/mol
Melting Pt: 282…286 °C
Storage Temperature: Ambient
MDL Number: MFCD04115756
CAS Number: 236388-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR