5,7-Difluoroquinazolin-4(3H)-one 95+%

Supplier: Apollo Scientific

PC201242-1G
APOSPC201242-1GEA 85.7 EUR
APOSPC201242-1G
5,7-Difluoroquinazolin-4(3H)-one 95+%
5,7-Difluoroquinazolin-4(3H)-one
Formula: C₈H₄F₂N₂O
MW: 182.1270 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100783
CAS Number: 379228-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR