2-Fluoroisonicotinic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Fluoropyridine-4-carboxylic acid

PC2057-1G
APOSPC2057-1GEA 21.9 EUR
APOSPC2057-1G
2-Fluoroisonicotinic acid 98%
2-Fluoroisonicotinic acid
Formula: C₆H₄FNO₂
MW: 141.10 g/mol
Melting Pt: 191…200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02181194
CAS Number: 402-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR