3-Bromo-2,2,3-trifluoropropanoic acid 97%

Supplier: Apollo Scientific
Danger

Synonyms: 3-Bromo-3H-perfluoropropanoic acid

PC2483-100MG
APOSPC2483-100MGEA 183 EUR
APOSPC2483-100MG
3-Bromo-2,2,3-trifluoropropanoic acid 97%
3-Bromo-2,2,3-trifluoropropanoic acid
Formula: C₃H₂BrF₃O₂
MW: 206.946 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412190
CAS Number: 679-95-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR