2-Bromo-2',4'-difluoroacetophenone 98%

Supplier: Apollo Scientific
Danger

Synonyms: 2-Bromo-1-(2,4-difluorophenyl)ethanone, 2,4-Difluorophenacyl bromide

PC2755-25G PC2755-5G PC2755-100G
APOSPC2755-25GEA 34.8 EUR
APOSPC2755-25G APOSPC2755-5G APOSPC2755-100G
2-Bromo-2',4'-difluoroacetophenone 98%
2-Bromo-2',4'-difluoroacetophenone
Formula: C₈H₅BrF₂O
MW: 235.03 g/mol
Melting Pt: 31…35 °C
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00142822
CAS Number: 102429-07-2
UN: 2925
ADR: 4.1+8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR