5-Fluoro-m-anisaldehyde 97+%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Fluoro-5-methoxybenzaldehyde, 5-Fluoro-3-methoxybenzaldehyde, 3-Fluoro-5-anisaldehyde

PC2772-1G PC2772-25G PC2772-5G
APOSPC2772-1GEA 21.9 EUR
APOSPC2772-1G APOSPC2772-25G APOSPC2772-5G
5-Fluoro-m-anisaldehyde 97+%
5-Fluoro-m-anisaldehyde
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 24…29 °C
Storage Temperature: Ambient
MDL Number: MFCD08741376
CAS Number: 699016-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR