Perfluoro-15-crown-5-ether 95%

Supplier: Apollo Scientific
Warning

Synonyms: Icosafluoro-15-crown 5-Ether, Icosafluoro-1,4,7,10,13-pentaoxacyclopentadecane, Perfluoro-1,4,7,10,13-pentaoxacyclopentadecane, Perfluoro-15-crown 5-Ether

PC3005-1G PC3005-5G PC3005-1KG PC3005-250G PC3005-500G PC3005-750G
APOSPC3005-1GEA 79.9 EUR
APOSPC3005-1G APOSPC3005-5G APOSPC3005-1KG APOSPC3005-250G APOSPC3005-500G APOSPC3005-750G
Perfluoro-15-crown-5-ether 95%
Perfluoro-15-crown-5-ether

Potential 19F NMR imaging agent, particularly effective in brain and spinal diagnostics, USP 4,838,274. Single nmr signal; liquid at room temperature.

Formula: C₁₀F₂₀O₅
MW: 580.08 g/mol
Boiling Pt: 146 °C
Density: 1.3 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01320747
CAS Number: 97571-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR