3-(Bromomethyl)-2-methyl-6-(trifluoromethyl)pyridine

Supplier: Apollo Scientific
Danger

PC300602-250MG
APOSPC300602-250MGEA 98.6 EUR
APOSPC300602-250MG
3-(Bromomethyl)-2-methyl-6-(trifluoromethyl)pyridine
3-(Bromomethyl)-2-methyl-6-(trifluoromethyl)pyridine
Formula: C₈H₇BrF₃N
MW: 254.047 g/mol
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD12923135
CAS Number: 917396-30-6
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR