5-(Trifluoromethyl)pyridin-3-amine

Supplier: Apollo Scientific
Warning

Synonyms: 3-Amino-5-(trifluoromethyl)pyridine

PC301200-1G PC301200-5G PC301200-25G
APOSPC301200-1GEA 29 EUR
APOSPC301200-1G APOSPC301200-5G APOSPC301200-25G
5-(Trifluoromethyl)pyridin-3-amine
5-(Trifluoromethyl)pyridin-3-amine
Formula: C₆H₅F₃N₂
MW: 162.1100 g/mol
Boiling Pt: 105.5…106 °C
Melting Pt: 40…41.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00128900
CAS Number: 112110-07-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR