1,4-Dibromo-2,3-difluorobenzene 97%

Supplier: Apollo Scientific
Warning

PC32740-5G PC32740-1G PC32740-25G
APOSPC32740-5GEA 27.7 EUR
APOSPC32740-5G APOSPC32740-1G APOSPC32740-25G
1,4-Dibromo-2,3-difluorobenzene 97%
1,4-Dibromo-2,3-difluorobenzene
Formula: C₆H₂Br₂F₂
MW: 271.8849 g/mol
Boiling Pt: 93…94 °C/50mmHg
Storage Temperature: Ambient
MDL Number: MFCD09835192
CAS Number: 156682-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR