Fluoroacetone 97%

Supplier: Apollo Scientific
Danger

PC3356-1G
APOSPC3356-1GEA 62.5 EUR
APOSPC3356-1G
Fluoroacetone 97%
Fluoroacetone
Formula: C₃H₅FO
MW: 76.07 g/mol
Boiling Pt: 75 °C
Density: 1.054 g/cm³ (25 °C)
Flash Pt: 7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000451
CAS Number: 430-51-3
EINECS: 207-064-0
UN: 2929
ADR: 6.1+3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR