3-Fluoroanisole 99%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Fluoro-3-methoxybenzene

PC3430-100G PC3430-25G PC3430-50G PC3430-1KG
APOSPC3430-100GEA 65.5 EUR
APOSPC3430-100G APOSPC3430-25G APOSPC3430-50G APOSPC3430-1KG
3-Fluoroanisole 99%
3-Fluoroanisole
Formula: C₇H₇FO
MW: 126.13 g/mol
Boiling Pt: 158…159 °C
Melting Pt: –35 °C
Density: 1.104 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00000335
CAS Number: 456-49-5
EINECS: 207-267-4
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR