2-Fluorobenzoic acid 99%

Supplier: Apollo Scientific
Warning

Synonyms: o-Fluorobenzoic acid

PC3500-1KG PC3500-100G PC3500-500G
APOSPC3500-1KGEA 55.1 EUR
APOSPC3500-1KG APOSPC3500-100G APOSPC3500-500G
2-Fluorobenzoic acid 99%
2-Fluorobenzoic acid
Formula: C₇H₅FO₂
MW: 140.11 g/mol
Boiling Pt: 245
Melting Pt: 122…125 °C
Density: 1.46 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002405
CAS Number: 445-29-4
EINECS: 207-158-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR