Bis(3,3,3-trifluoroprop-1-yl)amine 95%

Supplier: Apollo Scientific
Danger

Synonyms: 3,3,3-Trifluoro-N-(3,3,3-trifluoroprop-1-yl)propylamine

PC3539-1G PC3539-5G
APOSPC3539-1GEA 180 EUR
APOSPC3539-1G APOSPC3539-5G
Bis(3,3,3-trifluoroprop-1-yl)amine 95%
Bis(3,3,3-trifluoroprop-1-yl)amine
Formula: C₆H₉F₆N
MW: 209.133 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07784275
CAS Number: 201057-04-7
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR