2-Fluoro-6-(4-fluorophenoxy)benzonitrile 97%

Supplier: Apollo Scientific
Danger

PC3752D-250MG
APOSPC3752D-250MGEA 157 EUR
APOSPC3752D-250MG
2-Fluoro-6-(4-fluorophenoxy)benzonitrile 97%
2-Fluoro-6-(4-fluorophenoxy)benzonitrile
Formula: C₁₃H₇F₂NO
Melting Pt: 73…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00068202
CAS Number: 175204-07-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR