3-Fluorosalicylaldehyde 95%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Fluoro-2-hydroxybenzaldehyde

PC3756A-25G PC3756A-5G PC3756A-100G
APOSPC3756A-25GEA 125 EUR
APOSPC3756A-25G APOSPC3756A-5G APOSPC3756A-100G
3-Fluorosalicylaldehyde 95%
3-Fluorosalicylaldehyde
Formula: C₇H₅FO₂
MW: 140.12 g/mol
Boiling Pt: 80…85 °C/7mmHg
Melting Pt: 67…71 °C
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00003319
CAS Number: 394-50-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR