5-Fluoro-2-nitrobenzaldehyde 98%

Supplier: Apollo Scientific
Warning

PC3885D-100G PC3885D-250G PC3885D-25G PC3885D-500G
APOSPC3885D-100GEA 108 EUR
APOSPC3885D-100G APOSPC3885D-250G APOSPC3885D-25G APOSPC3885D-500G
5-Fluoro-2-nitrobenzaldehyde 98%
5-Fluoro-2-nitrobenzaldehyde
Formula: C₇H₄FNO₃
MW: 169.11 g/mol
Boiling Pt: 153 °C/23mmHg
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00153175
CAS Number: 395-81-3
EINECS: 206-903-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR