1-Fluoro-2-nitrobenzene 98%

Supplier: Apollo Scientific
Danger

Synonyms: o-Fluoronitrobenzene, 2-Fluoronitrobenzene

PC3890-1KG PC3890-250G PC3890-25G
APOSPC3890-1KGEA 127 EUR
APOSPC3890-1KG APOSPC3890-250G APOSPC3890-25G
1-Fluoro-2-nitrobenzene 98%
1-Fluoro-2-nitrobenzene

The activated fluorine reacts with arylamines to form 2-nitrodiarylamines see: Synthesis, 215 (1980)

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 116 °C/22mmHg
Melting Pt: –6 °C
Density: 1.338 g/cm³ (25 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00007048
CAS Number: 1493-27-2
EINECS: 216-088-0
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR