7-Fluoro-2-mercaptoquinazolin-4(3H)-one 95%

Supplier: Apollo Scientific
Warning

PC400047-1G
APOSPC400047-1GEA 192 EUR
APOSPC400047-1G
7-Fluoro-2-mercaptoquinazolin-4(3H)-one 95%
7-Fluoro-2-mercaptoquinazolin-4(3H)-one
Formula: C₈H₅FN₂OS
MW: 196.202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24573053
CAS Number: 69661-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR