4-(4-Trifluoromethylpyridin-2-yl)benzoic acid

Supplier: Apollo Scientific

PC400664-1G PC400664-250MG PC400664-500MG
APOSPC400664-1GEA 295 EUR
APOSPC400664-1G APOSPC400664-250MG APOSPC400664-500MG
4-(4-Trifluoromethylpyridin-2-yl)benzoic acid
4-(4-Trifluoromethylpyridin-2-yl)benzoic acid
Formula: C₁₃H₈F₃NO₂
MW: 267.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384864
CAS Number: 1299607-71-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR