Methyl 7-fluoro-1H-indazole-4-carboxylate

Supplier: Apollo Scientific

PC402008-250MG
APOSPC402008-250MGEA 140 EUR
APOSPC402008-250MG
Methyl 7-fluoro-1H-indazole-4-carboxylate
Methyl 7-fluoro-1H-indazole-4-carboxylate
Formula: C₉H₇FN₂O₂
MW: 194.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21607513
CAS Number: 1079993-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR