1-[Amino(4-fluorophenyl)methyl]naphthalen-2-ol

Supplier: Apollo Scientific

PC404506-1G PC404506-250MG
APOSPC404506-1GEA 0 EUR
APOSPC404506-1G APOSPC404506-250MG
1-[Amino(4-fluorophenyl)methyl]naphthalen-2-ol
1-[Amino(4-fluorophenyl)methyl]naphthalen-2-ol
Formula: C₁₇H₁₄FNO
Storage Temperature: Ambient
MDL Number: MFCD09260389
CAS Number: 714953-86-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR