ω-Bromo-4-fluoroacetophenone 97%

Supplier: Apollo Scientific
Danger

Synonyms: Bromomethyl p-fluorophenyl ketone, α-Bromo-4-fluoroacetophenone, 2-Bromo-1-(4-fluorophenyl)ethan-1-one, 2-Bromo-4'-fluoroacetophenone, 4-Fluorophenacyl bromide, 2-Bromo-1-(4-fluorophenyl)ethanone

PC4060-25G PC4060-5G PC4060-100G
APOSPC4060-25GEA 43.5 EUR
APOSPC4060-25G APOSPC4060-5G APOSPC4060-100G
ω-Bromo-4-fluoroacetophenone 97%
ω-Bromo-4-fluoroacetophenone

Intermediate for preparation of 1,2-diarylcyclopentenolcyclooxygenase inhibitors: J.Med.Chem.,37,3878,1994.

Formula: C₈H₆BrFO
MW: 217.04 g/mol
Boiling Pt: 150 °C/12mmHg
Melting Pt: 45…49 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00040830
CAS Number: 403-29-2
EINECS: 206-955-1
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR