(1E)-N'-Hydroxy-3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanimidamide

Supplier: Apollo Scientific

PC410423-1G
APOSPC410423-1GEA 386 EUR
APOSPC410423-1G
(1E)-N'-Hydroxy-3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanimidamide
(1E)-N'-Hydroxy-3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanimidamide
Formula: C₉H₁₃F₃N₄O
MW: 250.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20491476
CAS Number: 1006353-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR