4-Fluorophenylhydrazinium chloride 97%

Supplier: Apollo Scientific
Warning

Synonyms: (4-Fluorphenyl)hydrazin HCl, 4-Fluorophenylhydrazine hydrochloride, 1-(4-Fluorophenyl)hydrazine hydrochloride, (4-Fluorophenyl)hydrazine hydrochloride

PC4156-100G PC4156-25G PC4156-500G
APOSPC4156-100GEA 34.8 EUR
APOSPC4156-100G APOSPC4156-25G APOSPC4156-500G
4-Fluorophenylhydrazinium chloride 97%
4-Fluorophenylhydrazinium chloride
Formula: C₆H₈ClFN₂
MW: 162.59 g/mol
Melting Pt: 265 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012942
CAS Number: 823-85-8
EINECS: 212-521-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR