2,2,3,3,4,4,4-Heptafluorobutyl 2,2,3,4,4,4-hexafluorobutyl carbonate

Supplier: Apollo Scientific

PC450330-25G
APOSPC450330-25GEA 1540 EUR
APOSPC450330-25G
2,2,3,3,4,4,4-Heptafluorobutyl 2,2,3,4,4,4-hexafluorobutyl carbonate
2,2,3,3,4,4,4-Heptafluorobutyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Formula: C₉H₅F₁₃O₃
MW: 408.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28334323
CAS Number: 1980048-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR