4'-Chloro-5-fluoro-1,1'-biphenyl-2-amine

Supplier: Apollo Scientific

Synonyms: 2-Amino-4'-chloro-5-fluorobiphenyl

PC48313-1G PC48313-100MG PC48313-250MG PC48313-500MG PC48313-2.5G PC48313-5G
APOSPC48313-1GEA 366 EUR
APOSPC48313-1G APOSPC48313-100MG APOSPC48313-250MG APOSPC48313-500MG APOSPC48313-2.5G APOSPC48313-5G
4'-Chloro-5-fluoro-1,1'-biphenyl-2-amine
4'-Chloro-5-fluoro-1,1'-biphenyl-2-amine
Formula: C₁₂H₉ClFN
MW: 221.66 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12824033
CAS Number: 188731-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR