3-Methoxy-5-methylbenzotrifluoride 95%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 3-methyl-5-(trifluoromethyl)phenyl ether, 1-Methoxy-3-methyl-5-(trifluoromethyl)benzene

PC48636-1G PC48636-250MG
APOSPC48636-1GEA 551 EUR
APOSPC48636-1G APOSPC48636-250MG
3-Methoxy-5-methylbenzotrifluoride 95%
3-Methoxy-5-methylbenzotrifluoride
Formula: C₉H₉F₃O
MW: 190.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185712
CAS Number: 1214385-12-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR