3-Chloro-2-(trifluoromethyl)phenol

Supplier: Apollo Scientific

PC500171-1G PC500171-250MG PC500171-5G
APOSPC500171-1GEA 243 EUR
APOSPC500171-1G APOSPC500171-250MG APOSPC500171-5G
3-Chloro-2-(trifluoromethyl)phenol
3-Chloro-2-(trifluoromethyl)phenol
Formula: C₇H₄ClF₃O
MW: 196.554 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17000111
CAS Number: 106877-34-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR