6-Bromo-7-fluorobenzo[b]furan-2-carboxylic acid

Supplier: Apollo Scientific
Warning

PC500193-1G PC500193-250MG
APOSPC500193-1GEA 300 EUR
APOSPC500193-1G APOSPC500193-250MG
6-Bromo-7-fluorobenzo[b]furan-2-carboxylic acid
6-Bromo-7-fluorobenzo[b]furan-2-carboxylic acid
Formula: C₉H₄BrFO₃
MW: 259.029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27664866
CAS Number: 1823358-68-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR