5-Bromo-3-nitro-2-(trifluoromethyl)pyridine 95%

Supplier: Apollo Scientific

PC50183-1G PC50183-250MG PC50183-5G
APOSPC50183-1GEA 521 EUR
APOSPC50183-1G APOSPC50183-250MG APOSPC50183-5G
5-Bromo-3-nitro-2-(trifluoromethyl)pyridine 95%
5-Bromo-3-nitro-2-(trifluoromethyl)pyridine
Formula: C₆H₂BrF₃N₂O₂
MW: 270.991 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18255263
CAS Number: 1211540-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR