4-Iodo-2-(trifluoromethyl)benzoic acid

Supplier: Apollo Scientific

PC502640-5G
APOSPC502640-5GEA 72.5 EUR
APOSPC502640-5G
4-Iodo-2-(trifluoromethyl)benzoic acid
4-Iodo-2-(trifluoromethyl)benzoic acid
Formula: C₈H₄F₃IO₂
MW: 316.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09881756
CAS Number: 954815-11-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR